Merck
Four new koumine metabolites in rat liver microsomes.
Journal of Asian natural products research (2013-01-18)
Lin Zhang, Lan Du, Wen-Wen Lv, Qing-Chun Zhao, Wei Hua, Ye An, Tao Guo, Li-Jun Wu
ABSTRAKT
Four new metabolites M-1 [1,2,18,19-tetradehydro-4-demethyl-3,17-epoxy-7,20(2H,19H)-cyclovobasan], M-2 [1,2,4,21,18,19-hexadehydro-4-demethyl-3,17-epoxy-7,20(2H,19H)-cyclovobasan], M-3 [1,2,18,19-tetradehydro-4-demethyl-4-formaldehyde-3,17-epoxy-7,20(2H,19H)-cyclovobasan], and M-4 [1,2,4,21,18,19-hexadehydro-4-demethyl-4-oxy-3,17-epoxy-7,20(2H,19H)-cyclovobasan] were isolated from the chloroform extract of koumine incubated with phenobarbital-treated rat liver microsomes. The structures of M-1, M-2, M-3, and M-4 were elucidated by spectroscopic methods including ESI-TOF-MS, 1D, and 2D NMR experiments. The metabolic pathway of koumine was proposed. The cytotoxic activities between koumine and its metabolites were also compared in the A549 cell line.