Merck
Ligand binding affinities of arctigenin and its demethylated metabolites to estrogen receptor alpha.
Molecules (Basel, Switzerland) (2013-01-18)
Jong-Sik Jin, Jong-Hyun Lee, Masao Hattori
ABSTRAKT
Phytoestrogens are defined as plant-derived compounds with estrogen-like activities according to their chemical structures and activities. Plant lignans are generally categorized as phytoestrogens. It was reported that (-)-arctigenin, the aglycone of arctiin, was demethylated to (-)-dihydroxyenterolactone (DHENL) by Eubacterium (E.) sp. ARC-2. Through stepwise demethylation, E. sp. ARC-2 produced six intermediates, three mono-desmethylarctigenins and three di-desmethylarctigenins. In the present study, ligand binding affinities of (-)-arctigenin and its seven metabolites, including DHENL, were investigated for an estrogen receptor alpha, and found that demethylated metabolites had stronger binding affinities than (-)-arctigenin using a ligand binding screen assay method. The IC(50) value of (2R,3R)-2-(4-hydroxy-3-methoxybenzyl)-3-(3,4-dihydroxybenzyl)-butyrolactone was 7.9 × 10⁻⁴ M.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
γ-Butyrolactone, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Hydroxybutanoic acid lactone, ≥98%, FCC, FG
Sigma-Aldrich
(−)-Arctigenin, ≥95% (LC/MS-ELSD)