Merck
Synthesis of carbazole-based selenaporphyrin via annulation.
Organic letters (2013-01-25)
Motoki Masuda, Chihiro Maeda, Naoki Yoshioka
ABSTRAKT
Cu(I)-mediated alkoxylation of doubly 1,3-butadiyne-bridged carbazole dimer 1, followed by acid-catalyzed cyclization, provided furan-bridged carbazole dimer 3, while annulation reaction of 1 with selenium in the presence of hydrazine monohydrate provided selenophene-bridged carbazole dimer 5a. Oxidation of isophlorin 5a afforded carbazole-based selenaporphyrin 5b, which possessed distinct aromaticity and produced intensified and red-shifted absorption bands in the near-IR region.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Copper(I) iodide, purum, ≥99.5%
Sigma-Aldrich
Copper(I) iodide, 99.999% trace metals basis
Sigma-Aldrich
Copper(I) iodide, 98%
Copper(I) iodide ChemBeads