Merck
[Clinical microbiological investigation of vancomycin intermediate Staphylococcus aureus during glycopeptide therapy].
Kansenshogaku zasshi. The Journal of the Japanese Association for Infectious Diseases (2013-02-02)
Hirofumi Toda, Toshihiro Yamaguchi, Takayuki Miyara, Akihiro Hisato, Tomohide Matsushima, Takahiro Shimada, Hiroyuki Sano, Kenichi Nakae, Toshinori Kamisako
ABSTRAKT
We isolated three strains of vancomycin intermediate Staphylococcus aureus (VISA) from a blood sample of a patient with infective endocarditis (VISA-1), postoperative pneumonia sputum (VISA-2), and pyogenic spondylitis blood sample (VISA-3). These VISA strains did not carry vanA, vanB, vanC1, or vanC2/C3 genes. Cell wall thickening was observed. VISA-1 and VISA-3 PFGE patterns showed the completely same pattern compared to the PFGE pattern of methicillin-resistant Staphylococcus aureus first isolated from patients 1 and 3. After 10 days on brain heart infusion agar, wall thickening in all three type of VISA was unchanged, but VISA-2 and VISA-3 reversed vancomycin susceptibility. The most suitable use of vancomycin in patients with MRSA infection thus appears to be in reducing the opportunity for cell wall thickening.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Teicoplanin