Merck
  • Alkaloids from Sri Lankan curry-leaf (Murraya koenigii) display melanogenesis inhibitory activity: structures of karapinchamines A and B.
Alkaloids from Sri Lankan curry-leaf (Murraya koenigii) display melanogenesis inhibitory activity: structures of karapinchamines A and B.
Bioorganic & medicinal chemistry (2013-02-05)
Seikou Nakamura, Souichi Nakashima, Yoshimi Oda, Nami Yokota, Katsuyoshi Fujimoto, Takahiro Matsumoto, Tomoe Ohta, Keiko Ogawa, Sayuri Maeda, Shino Nishida, Hisashi Matsuda, Masayuki Yoshikawa
ABSTRAKT
A methanolic extract and its ethyl acetate-soluble fraction from Sri Lankan curry-leaf, the leaves of Murraya koenigii, inhibited melanogenesis in theophylline-stimulated murine B16 melanoma 4A5 cells. Two new carbazole alkaloids, karapinchamines A and B, were isolated from the ethyl acetate-soluble fraction together with 12 known carbazole alkaloids. The structures of karapinchamines A and B were determined by physicochemical analyses. The principal alkaloid constituents were found to display potent melanogenesis inhibitory activity. The structural requirements of the carbazole alkaloids for melanogenesis inhibitory activity were discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%