Merck
1,2,3,4,6-Penta-O-galloyl-β-D-glucopyranose inhibits angiogenesis via inhibition of capillary morphogenesis gene 2.
Journal of medicinal chemistry (2013-02-12)
Lorna M Cryan, Lauren Bazinet, Kaiane A Habeshian, Shugeng Cao, Jon Clardy, Kenneth A Christensen, Michael S Rogers
ABSTRAKT
Capillary morphogenesis gene 2 (CMG2) is a transmembrane extracellular matrix binding protein that is also an anthrax toxin receptor. We have shown that high-affinity CMG2 binders can inhibit angiogenesis and tumor growth. We recently described a high-throughput FRET assay to identify CMG2 inhibitors. We now report the serendipitous discovery that PGG (1,2,3,4,6-penta-O-galloyl-β-D-glucopyranose) is a CMG2 inhibitor with antiangiogenic activity. PGG is a gallotannin produced by a variety of medicinal plants that exhibits a wide variety of antitumor and other activities. We find that PGG inhibits CMG2 with a submicromolar IC50 and it also inhibits the migration of human dermal microvascular endothelial cells at similar concentrations in vitro. Finally, oral or intraperitoneal administration of PGG inhibits angiogenesis in the mouse corneal micropocket assay in vivo. Together, these results suggest that a portion of the in vivo antitumor activity of PGG may be the result of antiangiogenic activity mediated by inhibition of CMG2.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Penta-O-galloyl-β-D-glucose hydrate, ≥96% (HPLC)
Sigma-Aldrich
Tannic acid, ACS reagent