Merck
Additive effect of alpha-tocopherol and ascorbic acid in combating ethanol-induced hepatic fibrosis.
Redox report : communications in free radical research (2013-02-12)
P Prathibha, S Rejitha, R Harikrishnan, S Syam Das, P A Abhilash, M Indira
ABSTRAKT
To investigate the efficacy of combined administration of alpha-tocopherol (AT) and ascorbic acid (AA) in reducing ethanol-induced hepatotoxicity. Rats were maintained for 90 days and grouped as follows: I-control rats, II-ethanol, III-alpha-tocopherol, IV-ethanol+alpha-tocopherol, V-AA, VI-ethanol+ascorbic acid, VII-alpha-tocopherol+ascorbic acid, VIII-ethanol+alpha-tocopherol+ascorbic acid. At the end of the experimental period, markers of hepatic function, oxidative stress, and the expression of markers of inflammation and fibrosis were assayed. The markers of hepatic function, lipid peroxidation products, protein carbonyls, and the expression of nuclear factor kappa B, tumor necrosis factor alpha, transforming growth factor beta 1, cytochrome P4502E1, and collagen Type I were elevated after ethanol administration. All these parameters were reduced in the ethanol group administered AT and AA in combination. The activities of antioxidant enzymes which were reduced by ethanol administration were enhanced on combined administration of AT and AA. The reduction in hepatic fibrosis was almost 20% more in AT and AA co-administered group compared with AT and AA alone treated groups. Combined administration of fat soluble AT and water soluble AA was beneficial against ethanol-induced hepatotoxicity. This may be due to their different subcellular localizations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
DL-α-Tocopherol acetate, ≥96% (HPLC)
Sigma-Aldrich
DL-α-Tocopherol acetate, tested according to Ph. Eur.
Supelco
DL-alpha-Tocopherol acetate, analytical standard
Supelco
Alpha Tocopheryl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
α-Tocopherol acetate, European Pharmacopoeia (EP) Reference Standard
All-rac-alpha-tocopheryl acetate for peak identification, European Pharmacopoeia (EP) Reference Standard