- Gold-catalysed cascade rearrangements of ynamide propargyl esters.
Gold-catalysed cascade rearrangements of ynamide propargyl esters.
Chemical communications (Cambridge, England) (2013-02-13)
Stephen J Heffernan, James M Beddoes, Mary F Mahon, Alan J Hennessy, David R Carbery
PMID23399826
ABSTRAKT
The Au(I)-catalysed rearrangement of propargylic esters formed from an ynamide has been studied. The reaction is facile, and when conducted in the presence of a reactive indole nucleophile, leads to a cascade process whereby γ-indolyl α-acyloxyenamides are formed in good yield and excellent E-stereoselectivity.