Merck
Heterometallic cerium(IV) perrhenate, permanganate, and molybdate complexes supported by the imidodiphosphinate ligand [N(i-Pr2PO)2]-.
Inorganic chemistry (2013-02-15)
Guo-Cang Wang, Herman H Y Sung, Feng-Rong Dai, Wai-Hang Chiu, Wai-Yeung Wong, Ian D Williams, Wa-Hung Leung
ABSTRAKT
Heterometallic cerium(IV) perrhenate, permanganate, and molybdate complexes containing the imidodiphosphinate ligand [N(i-Pr2PO)2](-) have been synthesized, and their reactivity was investigated. Treatment of Ce[N(i-Pr2PO)2]3Cl (1) with AgMO4 (M = Re, Mn) afforded Ce[N(i-Pr2PO)2]3(ReO4) (2) or Ce2[N(i-Pr2PO)2]6(MnO4)2 (3). In the solid state, 3 is composed of a [Ce2{N(i-Pr2PO)2}6(MnO4)](+) moiety featuring a weak Ce-OMn interaction [Ce-OMn distance = 2.528(8) Å] and a noncoordinating MnO4(-) counteranion. While 3 is stable in the solid state and acetonitrile solution, it decomposes readily in other organic solvents, such as CH2Cl2. 3 can oxidize ethylbenzene to acetophenone at room temperature. Treatment of 1 with AgBF4, followed by reaction with [n-Bu4N]2[MoO4], afforded [Ce{N(i-Pr2PO)2}3]2(μ-MoO4) (4). Reaction of trans-Ce[N(i-Pr2PO)2]2(NO3)2 (5), which was prepared from (NH4)2Ce(NO3)6 and K[N(i-Pr2PO)2], with 2 equiv of [n-Bu4N][Cp*MoO3] yielded trans-Ce[N(i-Pr2PO)2]2(Cp*MoO3)2 (6). 4 can catalyze the oxidation of methyl phenyl sulfide with tert-butyl hydroperoxide with high selectivity. The crystal structures of complexes 3-6 have been determined.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonium perrhenate, ≥99%
Sigma-Aldrich
Ammonium perrhenate, 99.999% trace metals basis
Sigma-Aldrich
Rhenium, powder, 99.995% trace metals basis
Sigma-Aldrich
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Potassium perrhenate, 99.98% trace metals basis
Sigma-Aldrich
Potassium perrhenate, 99%
Sigma-Aldrich
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Cerium, chips
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Sodium perrhenate, 99.99% trace metals basis