Merck
  • reliable Synthesis of Monodisperse Microparticles: Prevention of Oxygen Diffusion and Organic Solvents Using Conformal Polymeric Coating onto Poly(dimethylsiloxane) Micromold.
reliable Synthesis of Monodisperse Microparticles: Prevention of Oxygen Diffusion and Organic Solvents Using Conformal Polymeric Coating onto Poly(dimethylsiloxane) Micromold.
Langmuir : the ACS journal of surfaces and colloids (2013-02-15)
Jae-Min Jeong, Myung Seok Oh, Bong Jun Kim, Chang-Hyung Choi, Bora Lee, Chang-Soo Lee, Sung Gap Im
ABSTRAKT
An effective polymeric thin film deposited by initiated chemical vapor deposition (iCVD) process was presented and its application as a barrier film on the PDMS micromold blocking the penetration of oxygen and organic solvents was investigated. With this barrier film, we were able to synthesize monodisperse polymeric particles of sizes down to 3 μm, which has been reported to be extremely challenging with bare PDMS micromold. The polymeric barrier film on the PDMS micromold enabled this successful synthesis of microparticles by effectively blocking the diffusion of oxygen, which is a well-known radical quencher in radical polymerization, through the PDMS micromold. Furthermore, the iCVD barrier film substantially decreased the penetration of various organic solvents such as acetone, tert-butanol, PDMS oil, and decane as well as organic substances including fluorescent molecules like rhodamine B and fluorescein isothiocyanate (FITC). Therefore, the polymeric barrier film coated on PDMS micromold via iCVD process will broaden the application of PDMS to microfluidic area for the synthesis of smaller microparticles with various organic substances.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Supelco
Decane, analytical standard
Sigma-Aldrich
Decane, anhydrous, ≥99%
Sigma-Aldrich
Decane, ReagentPlus®, ≥99%
Sigma-Aldrich
Decane, ≥95%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
tert-Butanol, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butanol, TEBOL® 99, ≥99.3%
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
tert-Butanol, suitable for HPLC, ≥99.5%
Sigma-Aldrich
tert-Butanol, ≥99% (GC)
Supelco
tert-Butanol, analytical standard