Merck
Benchmarking for perturbative triple-excitations in EE-EOM-CC methods.
The journal of physical chemistry. A (2013-02-15)
Thomas J Watson, Victor F Lotrich, Peter G Szalay, Ajith Perera, Rodney J Bartlett
ABSTRAKT
Perturbative triples corrections ((T) and (T̃)) to the equation of motion coupled cluster singles and doubles (EOM-CCSD) are rederived and implemented in a pilot parallel code. The vertical excitation energies of molecules in the test set of Sauer et al. [J. Chem. Theor. Comput. 2009, 5, 555] are reported and compared to the iterative EOM-CCSDT-3 method. The average absolute deviations of EOM-CCSD(T) and EOM-CCSD(T̃) from EOM-CCSDT-3 over this wide test set are 0.06 and 0.18 eV, respectively. The poor performance of the latter suggests misbalanced handling of the (T̃) terms. Scaling curves for EOM-CCSD(T) are also presented to demonstrate the performance across multiple compute nodes, thus enabling the routine and accurate study of excited states for ever larger molecular systems.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thymine, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymine, ≥99%