Merck
Aspulvinones from a mangrove rhizosphere soil-derived fungus Aspergillus terreus Gwq-48 with anti-influenza A viral (H1N1) activity.
Bioorganic & medicinal chemistry letters (2013-02-16)
Huquan Gao, Wenqiang Guo, Qiang Wang, Lianqing Zhang, Meilin Zhu, Tianjiao Zhu, Qianqun Gu, Wei Wang, Dehai Li
ABSTRAKT
A new butenolide isoaspulvinone E (1), together with two known butenolides aspulvinone E (2) and pulvic acid (3) were isolated from the marine-derived fungus, Aspergillus terreus Gwq-48. They showed significant anti-influenza A H1N1 virus activities, with IC50 values of 32.3, 56.9, and 29.1μg/mL, respectively. Moreover, only compound 1 exhibited effective inhibitory activity against H1N1 viral neuraminidase (NA), and docking of two isomers (1-2) into the active sites of NA showed that the E double bond Δ(5(10)) was essential to achieve activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Hydroxybutanoic acid lactone, ≥98%, FCC, FG
Sigma-Aldrich
2(5H)-Furanone, 98%
Sigma-Aldrich
γ-Butyrolactone, ReagentPlus®, ≥99%