Merck
Stereoselective synthesis of multiple stereocenters by using a double aldol reaction.
Angewandte Chemie (International ed. in English) (2013-02-19)
Yasushi Shimoda, Tatsunori Kubo, Masaharu Sugiura, Shunsuke Kotani, Makoto Nakajima
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Supelco
Benzaldehyde, analytical standard