Merck
How safe is Celecoxib for Asian-Indian patients with rheumatic diseases?
International journal of rheumatic diseases (2013-02-28)
Debashish Danda, Mohamed M Iliyas, Sujith J Chandy, Cheemalapati Chandra, Ashish J Mathew
ABSTRAKT
Cyclo-oxygenase (COX)-2 inhibitors have been the target of severe criticism, more so following the withdrawal of Rofecoxib. Post-marketing surveillance of Celecoxib in Asian Indians, who are predisposed to premature athero-thrombotic events, has not been studied. To study the adverse effects of Celecoxib and compare them with those of other non-steroidal anti-inflammatory drugs (NSAIDs) in an Asian Indian cohort. This is a retrospective chart review with convenience sampling of patients on NSAIDs (at least five tablets a week, for at least 3 months prior to the study), attending the Rheumatology clinic of a tertiary care institution in south India between June 2004 and November 2004. Those with pre-existing heart disease, hypertension, thrombo-embolic disease, peptic ulcer and patients on corticosteroids were excluded. All the recorded adverse events were noted and compared between the Celecoxib and non-selective NSAID users. Univariate analysis using Chi-square test was performed. Of the 1387 patients included, 915 were on Celecoxib. In the NSAID group, 204 had used multiple NSAIDs in sequence. Of the Celecoxib users, 164 had switched over to an NSAID during the study period. New onset of hypertension was significantly higher in the Celecoxib users as compared to non-selective NSAID users (3.06% vs. 1.27%, P = 0.04). However, those who had switched over to NSAIDs did not show this trend. NSAID users, on the other hand, had significant gastrointestinal (GI) toxicity (2.54% vs. 0.327%, P = 0.001). A significant number of Celecoxib users who switched over to NSAIDs also developed GI toxicity (6.1% vs. 1.21%, P = 0.018) over a shorter time span, as compared to the continuous NSAID users. Multiple NSAID users had higher adverse events (6.37% vs. 2.23%, P = 0.023) as compared to single NSAID users. Celecoxib significantly increased the incidence of new onset hypertension in this cohort of Indian patients with rheumatic diseases. No thromboembolic events were documented.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Diclofenac sodium salt
Supelco
Diclofenac sodium salt, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Naproxen, meets USP testing specifications
Naproxen, European Pharmacopoeia (EP) Reference Standard
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Diclofenac sodium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Supelco
Naproxen, VETRANAL®, analytical standard
Supelco
Naproxen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®