Merck
Protective effect of butylated hydroxytoluene on ferric nitrilotriacetate induced hepatotoxicity and oxidative stress in mice.
Human & experimental toxicology (2013-02-28)
S Ansar, H Tabassum, N Al Jameil
ABSTRAKT
The present study was undertaken to evaluate the possible ameliorating effect of butylated hydroxyl toluene (BHT), associated with ferric nitrilotriacetate (Fe-NTA)-induced oxidative stress and liver injury in mice. The treatment of mice with Fe-NTA alone enhances ornithine decarboxylase activity to 4.6 folds, protein carbonyl formation increased up to 2.9 folds and DNA synthesis expressed in terms of [(3)H] thymidine incorporation increased to 3.2 folds, and antioxidants and antioxidant enzymes decreased to 1.8-2.5 folds, compared with the corresponding saline-treated controls. These changes were reversed significantly (p < 0.001) in animals receiving a pretreatment of BHT. Our data show that BHT can reciprocate the toxic effects of Fe-NTA and can serve as a potent chemopreventive agent.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Nitrilotriacetic acid, BioUltra, ≥99.0% (T)
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Supelco
3,5-Di-tert-butyl-4-hydroxytoluene, analytical standard
Butylhydroxytoluene, European Pharmacopoeia (EP) Reference Standard