Merck
Nitroso-redox imbalance affects cardiac structure and function.
Journal of the American College of Cardiology (2013-03-02)
Vasileios Karantalis, Ivonne Hernandez Schulman, Joshua M Hare
MATERIAŁY
Numer produktu
Marka
Opis produktu
Allopurinol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor