Merck
UVA-riboflavin collagen cross-linking: a misnomer perhaps, but it works!
Investigative ophthalmology & visual science (2013-03-07)
Stephen D Klyce
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, 70-79% (fluorimetric)
Sigma-Aldrich
Wodzian fosforanu sodu ryboflawiny, tested according to Ph. Eur.
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, synthetic, ≥70% (HPLC)
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, suitable for electrophoresis, suitable for acrylamide photopolymerization, ≥70%, powder
Riboflavin sodium phosphate, European Pharmacopoeia (EP) Reference Standard