Merck
  • A seeded synthetic strategy for uniform polymer and carbon nanospheres with tunable sizes for high performance electrochemical energy storage.
A seeded synthetic strategy for uniform polymer and carbon nanospheres with tunable sizes for high performance electrochemical energy storage.
Chemical communications (Cambridge, England) (2013-03-08)
Jiasheng Qian, Mingxian Liu, Lihua Gan, Pranav K Tripathi, Dazhang Zhu, Zijie Xu, Zhixian Hao, Longwu Chen, Dominic S Wright
ABSTRAKT
We established a novel and facile strategy to synthesize uniform polymer and carbon nanospheres, the diameters of which can be precisely programmed between 35-105 and 30-90 nm, respectively, via time-controlled formation of colloidal seeds. The carbon nanospheres show promising prospects in high rate performance electrochemical energy storage.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phloroglucinol, ≥99.0% (HPLC)
Sigma-Aldrich
Phloroglucinol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Phloroglucinol, Used to detect the presence of wood fiber.