Merck
  • Cyclization and N-iodosuccinimide-induced electrophilic iodocyclization of 3-aza-1,5-enynes to synthesize 1,2-dihydropyridines and 3-iodo-1,2-dihydropyridines.
Cyclization and N-iodosuccinimide-induced electrophilic iodocyclization of 3-aza-1,5-enynes to synthesize 1,2-dihydropyridines and 3-iodo-1,2-dihydropyridines.
The Journal of organic chemistry (2013-03-12)
Xiaoyi Xin, Dongping Wang, Fan Wu, Xincheng Li, Boshun Wan
ABSTRAKT
Metal-free cyclization and N-iodosuccinimide-induced electrophilic iodocyclization of readily available 3-aza-1,5-enynes have been developed. The reactions selectively give 1,2-dihydropyridines and 3-iodo-1,2-dihydropyridines involving an aza-Claisen rearrangement and a 6π-electrocyclization step. Furthermore, the reaction could be carried out in 10 g scale for the synthesis of 1,2-dihydropyridines.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-Iodosuccinimide, 95%