Merck
High-throughput fluorescence assay of cytochrome P450 3A4.
Methods in molecular biology (Clifton, N.J.) (2013-03-12)
Qian Cheng, F Peter Guengerich
ABSTRAKT
Microtiter plate-based fluorescence assays allow rapid measurement of the catalytic activities of cytochrome P450 oxygenases (P450s). We describe a high-throughput fluorescence assay of P450 3A4, one of the key enzymes involved in xenobiotic metabolism. The assay involves the oxidative debenzylation of 7-hydroxy-4-trifluoromethyl coumarin, producing an increase in fluorescence.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Coumarin, primary reference standard
Supelco
Coumarin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland