Merck
Encodable multiple-fluorescence CdTe@carbon nanoparticles from nanocrystal/colloidal crystal guest-host ensembles.
Nanotechnology (2013-03-13)
Xin Guo, Cai-Feng Wang, Li-Hua Mao, Jing Zhang, Zi-Yi Yu, Su Chen
ABSTRAKT
We report herein the controllable generation of encodable multi-fluorescence CdTe@carbon nanoparticles (CdTe@C NPs) via the pyrolysis of quantum dot/photonic crystal (QD/PC) guest-host ensembles. The precursors of CdTe/poly(styrene-co-glycidylmethacrylate) (PS-co-PGMA) QD/PC guest-host ensembles were initially formed via the assembly of epoxy groups of PCs and carboxyl groups on the surface of CdTe QDs, followed by a pyrolysis process to generate CdTe@C NPs. The as-prepared CdTe@C NPs not only integrate the optical properties for both the carbon and CdTe QD constituents, but also enable an impressive enhancement of the fluorescence lifetime for CdTe QDs. The multifarious fluorescent spectra coding for CdTe@C NPs was further generated through regulating the embedded sizes or concentrations of CdTe QDs and the excitation wavelength, and their applications in DNA detection and luminescent patterns were achieved.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tellurium, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, powder, −30 mesh, 99.997% trace metals basis
Sigma-Aldrich
Tellurium, granular, −5-+50 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tellurium, powder, −200 mesh, 99.8% trace metals basis
Sigma-Aldrich
Tellurium, shot, ≤2 mm, 99.999% trace metals basis