- Stability of isoenzymes of alkaline phosphatase in various buffer systems.
Stability of isoenzymes of alkaline phosphatase in various buffer systems.
Enzyme (1979-01-01)
K Jung, M Pergande, E Egger
PMID436806
ABSTRAKT
The stability of isoenzymes of alkaline phosphatase from liver, bones and small intestine was compared after addition to inactivated serum in the buffer systems: glycine, 2-amino-2-methyl-1-propanol, diethanolamine and 2-amino-2-methyl-1,3-propandiol at 37 degrees C. The mentioned isoenzymes were inactivated to different extents in glycine and 2-amino-2-methyl-1-propanol buffers. In diethanolamine and 2-amino-2-methyl-1,3-propandiol buffers sufficient stability of isoenzymes is obtained so that only these buffers are suitable for activity determinations of alkaline phosphatase at 37 degrees C.