Merck
Study of removal of Direct Yellow 12 by cadmium oxide nanowires loaded on activated carbon.
Materials science & engineering. C, Materials for biological applications (2013-03-19)
Mehrorang Ghaedi, Batuol Sadeghian, Syamak Nasiri Kokhdan, Arezou Amiri Pebdani, Reza Sahraei, Ali Daneshfar, Asma Mihandoost
ABSTRAKT
In this research, cadmium oxide nanowires loaded on activated carbon (CdO-NW-AC) has been synthesized by a simple procedure and characterized by different techniques such as XRD, SEM and UV-vis spectrometry. This new adsorbent has been efficiently utilized for the removal of the Direct Yellow 12 (DY-12) from wastewater. To obtain maximum DY-12 removal efficiency, the influences of variables such as pH, DY-12 concentration, amount of CdO-NW-AC, contact time, and temperature have been examined and optimized in a batch method. Following the variable optimization, the experimental equilibrium data (at different concentration of DY-12) was fitted to conventional isotherm models such as Langmuir, Freundlich and Tempkin. The applicability of each method is based on the R(2) and error analysis for each model. It was found that the experimental equilibrium data well fitted to the Langmuir isotherm model. The dependency of removal process to time and the experimental data follow second order kinetic model with involvement of intraparticle diffusion model. The negative value of Gibbs's free energy and positive value of adsorption enthalpy show the spontaneous and endothermic nature of adsorption process.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cadmium oxide, powder, 99.5% trace metals basis
Sigma-Aldrich
Cadmium oxide, ≥99.99% trace metals basis