- Development of a "signal-on" electrochemical DNA sensor with an oligo-thymine spacer for point mutation detection.
Development of a "signal-on" electrochemical DNA sensor with an oligo-thymine spacer for point mutation detection.
Chemical communications (Cambridge, England) (2013-03-19)
Yao Wu, Rebecca Y Lai
PMID23503676
ABSTRAKT
We report a reagentless and reusable "signal-on" electrochemical DNA sensor that utilizes oligo-thymine as a flexible spacer. This sensor design is sensitive, specific, and selective; it is potentially generalizable for detection of a wide range of biomedical-relevant nucleic acid sequences.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution