- Should insulin resistance degree be taken into account for assessment of glycemic index?
Should insulin resistance degree be taken into account for assessment of glycemic index?
The American journal of clinical nutrition (2013-03-22)
Jose E Galgani, Giovanna Valentino
PMID23515393
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20