Merck
The histone demethylase PHF8 governs retinoic acid response in acute promyelocytic leukemia.
Cancer cell (2013-03-23)
Maria Francisca Arteaga, Jan-Henrik Mikesch, Jihui Qiu, Jesper Christensen, Kristian Helin, Scott C Kogan, Shuo Dong, Chi Wai Eric So
ABSTRAKT
While all-trans retinoic acid (ATRA) treatment in acute promyelocytic leukemia (APL) has been the paradigm of targeted therapy for oncogenic transcription factors, the underlying mechanisms remain largely unknown, and a significant number of patients still relapse and become ATRA resistant. We identified the histone demethylase PHF8 as a coactivator that is specifically recruited by RARα fusions to activate expression of their downstream targets upon ATRA treatment. Forced expression of PHF8 resensitizes ATRA-resistant APL cells, whereas its downregulation confers resistance. ATRA sensitivity depends on the enzymatic activity and phosphorylation status of PHF8, which can be pharmacologically manipulated to resurrect ATRA sensitivity to resistant cells. These findings provide important molecular insights into ATRA response and a promising avenue for overcoming ATRA resistance.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)