- Indium/TFA-catalyzed synthesis of tetracyclic [6,5,5,6] indole ring, via a tandem cycloannulation of β-oxodithioester with tryptamine.
Indium/TFA-catalyzed synthesis of tetracyclic [6,5,5,6] indole ring, via a tandem cycloannulation of β-oxodithioester with tryptamine.
Organic letters (2013-04-12)
Thokchom Prasanta Singh, Suman Bhattarcharya, Okram Mukherjee Singh
PMID23573995
ABSTRAKT
The cycloannulation of β-oxodithioesters and tryptamine in dichloromethane in the presence of a catalytic amount of InCl3 and TFA gave the novel 5-aryl/heteroaryl 2a(1),9b-dihydro-1H-2a,5a-diaza-cyclopenta[jk]fluorene-3(2H)-thiones in moderate to good yields. The reaction was proposed to involve a tandem transformation of thioamide, protonation, and dehydrative cyclization. KMnO4-oxidation of these newly prepared compounds yielded oxidative desulfurization products in good yields.