Merck
Fundus autofluorescence in hydroxychloroquine toxicity.
JAMA ophthalmology (2013-04-13)
Ian R Gorovoy, Mark S Gorovoy
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydroxychloroquine sulfate, ≥98% (HPLC), powder