Merck
Does increased red blood cell deformability raise the risk for osteonecrosis in sickle cell anemia?
Blood (2013-04-13)
Nathalie Lemonne, Yann Lamarre, Marc Romana, Martin Mukisi-Mukaza, Marie-Dominique Hardy-Dessources, Vanessa Tarer, Danielle Mougenel, Xavier Waltz, Benoît Tressières, Marie-Laure Lalanne-Mistrih, Maryse Etienne-Julan, Philippe Connes
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydroxyurea, 98%, powder