Merck
BTEX pollution caused by motorcycles in the megacity of HoChiMinh.
Journal of environmental sciences (China) (2013-04-20)
Tran Thi Ngoc Lan, Pham Anh Minh
ABSTRAKT
Monitoring of benzene, toluene and xylenes (BTEX) was conducted along with traffic counts at 17 roadside sites in urban areas of HoChiMinh. Toluene was the most abundant substance, followed by p,m-xylenes, benzene, o-xylene and ethylbenzene. The maximum observed hour-average benzene concentration was 254 microg/m3. Motorcycles contributed to 91% of the traffic fleet. High correlations among BTEX species, between BTEX concentrations and the volume of on-road motorcycles, and between inter-species ratios in air and in gasoline indicate the motorcycle-exhaust origin of BTEX species. Daily concentrations of benzene, toluene, ethylbenzene, p,m-xylenes and o-xylene were 56, 121, 21, 64 and 23 microg/m3, respectively, p,m-xylenes possess the highest ozone formation potential among the BTEX family.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
Supelco
Toluene, analytical standard
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Supelco
Benzene, analytical standard
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%