Merck
Free-breathing, motion-corrected late gadolinium enhancement is robust and extends risk stratification to vulnerable patients.
Circulation. Cardiovascular imaging (2013-04-20)
Kayla M Piehler, Timothy C Wong, Kathy S Puntil, Karolina M Zareba, Kathie Lin, David M Harris, Christopher R Deible, Joan M Lacomis, Ferenc Czeyda-Pommersheim, Stephen C Cook, Peter Kellman, Erik B Schelbert
ABSTRAKT
Routine clinical use of novel free-breathing, motion-corrected, averaged late-gadolinium-enhancement (moco-LGE) cardiovascular MR may have advantages over conventional breath-held LGE (bh-LGE), especially in vulnerable patients. In 390 consecutive patients, we collected bh-LGE and moco-LGE with identical image matrix parameters. In 41 patients, bh-LGE was abandoned because of image quality issues, including 10 with myocardial infarction. When both were acquired, myocardial infarction detection was similar (McNemar test, P=0.4) with high agreement (κ=0.95). With artifact-free bh-LGE images, pixelwise myocardial infarction measures correlated highly (R(2)=0.96) without bias. Moco-LGE was faster, and image quality and diagnostic confidence were higher on blinded review (P<0.001 for all). During a median of 1.2 years, 20 heart failure hospitalizations and 18 deaths occurred. For bh-LGE, but not moco-LGE, inferior image quality and bh-LGE nonacquisition were linked to patient vulnerability confirmed by adverse outcomes (log-rank P<0.001). Moco-LGE significantly stratified risk in the full cohort (log-rank P<0.001), but bh-LGE did not (log-rank P=0.056) because a significant number of vulnerable patients did not receive bh-LGE (because of arrhythmia or inability to hold breath). Myocardial infarction detection and quantification are similar between moco-LGE and bh-LGE when bh-LGE can be acquired well, but bh-LGE quality deteriorates with patient vulnerability. Acquisition time, image quality, diagnostic confidence, and the number of successfully scanned patients are superior with moco-LGE, which extends LGE-based risk stratification to include patients with vulnerability confirmed by outcomes. Moco-LGE may be suitable for routine clinical use.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Gadolinium, chips
Sigma-Aldrich
Gadolinium, −40 mesh, 99% trace rare earth metals basis