Merck
[Fatal zygomycosis caused by Mucor indicus after haplo-identical stem cell transplantation].
[Rinsho ketsueki] The Japanese journal of clinical hematology (2013-05-17)
Satoko Koteda, Kei Nomura, Michitoshi Hashiguchi, Kuniki Kawaguchi, Eijiro Oku, Koichi Osaki, Takayuki Nakamura, Fumihiko Mouri, Rie Imamura, Ritsuko Seki, Koji Nagafuji, Koichi Makimura, Takashi Okamura
ABSTRAKT
A 62-year-old woman with acute lymphoblastic leukemia in first complete remission was treated with unrelated cord blood transplantation, but exhibited primary graft failure. She then underwent HLA-haploidentical peripheral blood stem cell transplantation from her daughter. The conditioning regimen consisted of fludarabine 30 mg/m(2)/day for 6 days, intravenous busulfan 3.2 mg/kg/day for 2 days, and thymoglobulin 1 mg/kg/day for 2 days. Voriconazole was administered to prevent fungal infections. The patient achieved prompt hematopoietic recovery. Fever was observed 21 days after the second transplant, followed by sigmoid colon perforation and a liver space occupying lesion (SOL). A filamentous fungus was detected in a percutaneous biopsy of the liver SOL. In spite of changing the antifungal drug from voriconazole to liposomal amphotericin B, the patient died on day 41. The fungus was identified as Mucor indicus, a type of zygomycete. Although Mucor indicus inhabits soil, an infectious disease is extremely rare, and breakthrough infection after voriconazole prophylaxis had not been reported until now. It is mandatory to consider preventive antifungal treatment for drug-resistant fungal infectious diseases in patients after neutrophilic recovery with a strongly immunocompromised state after a HLA-haploidentical transplant.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Fluoroadenine-9-β-D-arabinofuranoside, DNA synthesis and methylation inhibitor
Sigma-Aldrich
Adenine 9-β-D-arabinofuranoside, ≥99%