Merck
Anomalous Schottky specific heat and structural distortion in ferromagnetic PrAl2.
Physical review letters (2013-05-21)
Arjun K Pathak, D Paudyal, Y Mudryk, K A Gschneidner, V K Pecharsky
ABSTRAKT
Unique from other rare earth dialuminides, PrAl(2) undergoes a cubic to tetragonal distortion below T = 30 K in a zero magnetic field, but the system recovers its cubic symmetry upon the application of an external magnetic field of 10 kOe via a lifting of the 4f crystal field splitting. The nuclear Schottky specific heat in PrAl(2) is anomalously high compared to that of pure Pr metal. First principles calculations reveal that the 4f crystal field splitting in the tetragonally distorted phase of PrAl(2) underpins the observed unusual low temperature phenomena.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Aluminum, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis