Merck
Neutralizing a caustic chemical.
Nursing (2013-05-21)
Santiago Nogué, Montserrat Amigó
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis