Merck
[Statement of the Polish Gynecological Society expert panel concerning application of Fluomizin].
Ginekologia polska (2013-05-25)
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Dequalinium chloride, Pharmaceutical Secondary Standard; Certified Reference Material