Merck
Pharmacokinetic evaluation of hydrocodone/acetaminophen for pain management.
Journal of opioid management (2013-05-28)
Aarti Singla, Paul Sloan
ABSTRAKT
Hydrocodone/acetaminophen is not only the most commonly prescribed opioid in the United States but also the most common prescription medication written in America. Although original and early trials confirmed its ability to manage acute pain from surgery and musculoskeletal injury, it is perhaps more widely used today in the management of chronic pain. However, the opioid product was introduced for the management of moderate to moderately severe pain. Because it has been greatly abused as a prescription opioid medication, physicians need to be aware of the current knowledge regarding this analgesic drug. This review summarizes the current knowledge of the pharmacokinetics, pharmacodynamics, and metabolism of hydrocodone. Recent information regarding the possibility of hydrocodone as a prodrug for hydromorphone is discussed. The available clinical trials for the use of hydrocodone in the management of acute, chronic, and cancer pain are presented.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrocodone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Acetaminofen, United States Pharmacopeia (USP) Reference Standard