Merck
Editorial: A missing link? Monocyte activation by uremic toxins in cardiorenal syndrome.
Journal of leukocyte biology (2013-06-01)
Ehrin J Armstrong, Jennifer L Granick, Scott I Simon
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Indoxyl sulfate potassium salt