Merck
Weekly paclitaxel--an effective treatment for advanced breast cancer.
Anticancer research (2013-06-12)
Pirkko Kellokumpu-Lehtinen, Tuija Tuunanen, Raija Asola, Liisa Elomaa, Mirja Heikkinen, Riitta Kokko, Ritva Järvenpää, Ilari Lehtinen, Abdel Maiche, Jaana Kaleva-Kerola, Mauri Huusko, Kari Möykkynen, Timo Ala-Luhtala
ABSTRAKT
Weekly paclitaxel is widely used in the treatment of metastatic breast cancer (MBC). Our aim was to test its efficacy and tolerability as a second-line therapy for MBC in daily oncology practice. Paclitaxel (90 mg/m(2)) was given intravenously three times weekly in a 4-week cycle to 91 patients with disease progression after hormonal (42%) or cytostatic therapy (57%). The median age was 54 years; metastatic sites were the lung (39%), liver (52%) and bone (47%). 64% of patients had more than one site of metastasis. Median time-to-progression was 7.5 months (range=6.5-8.5 months) and median overall survival time was 20.1 months (range=13.7-26.5 months). We observed 10 complete (12%) and 37 partial (43%) responses (an overall response rate of 55%). Severe side-effects were rare (grade 3-4 neutropenia 13% and septic episodes in three cases). Weekly paclitaxel was shown to be an effective and well-tolerated treatment for advanced breast cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Paclitaxel, European Pharmacopoeia (EP) Reference Standard