Merck
The role of Dr Isaac Aaron and the Australian Medical Journal in the dissemination of information about etherisation in the 1840s.
Anaesthesia and intensive care (2013-07-24)
J D Paull
ABSTRAKT
Isaac Aaron (1804 to 1877), an ambitious young medical practitioner, arrived in Sydney from Britain in 1838 and was registered by the New South Wales Medical Board the following year. After contributing to the Australian Medical Journal, established in August 1846 by William Baker, he became the editor in December and acquired it in May the following year. Dr Pugh became the most prolific local contributor to the journal but he and the editor had a somewhat 'prickly' relationship. Aaron was very critical of etherisation when the first news arrived in Australia, but Pugh chose Aaron's journal in which to report his initial and subsequent experience with the technique. Aaron repeatedly appealed for experimental evidence and rational decision-making to determine the place of etherisation in medical practice. Unfortunately for Australian medicine, Aaron had to suspend the publication of the journal in October 1847, lacking both time and the support of the profession necessary to maintain it. This created an unanticipated adverse outcome for Dr Pugh.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor