Merck
Anti-inflammatory and gastroprotective activities of the aqueous extract of Micromeria fruticosa (L.) Druce ssp Serpyllifolia in mice.
Pakistan journal of pharmaceutical sciences (2013-07-03)
Eman Abu-Gharbieh, Naglaa Gamil Shehab, Saeed Ahmed Khan
ABSTRAKT
Micromeria fruticosa is used widely in many Mediterranean regions for various inflammatory conditions. The aim of this work was to assess the anti-inflammatory and gastroprotective activities of the aqueous extract of Micromeria fruticosa. The aqueous extract of Micromeria fruticosa was tested orally in mice at doses of 50, 100 and 200 mg/kg in carrageenan-induced paw edema, vascular permeability, myeloperoxidase activity (MPO) and indomethacin-induced gastric ulceration. In the paw edema model, the extract at dose of 200 mg/ kg, exhibited a significant anti-inflammatory effect, while the extract at 100 and 200 mg/kg reduced significantly the vascular permeability and MPO activity in a dose dependant manner. Oral pretreatment of the aqueous extract reduced significantly the development of gastric lesions induced by indomethacin at dose of 200 mg/kg only. Results suggest that the aqueous extract of Micromeria fruticosa has both anti-inflammatory as well as, gastroprotective activities. Thus it could be used as an alternative or supplementary herbal remedy for the treatment of inflammatory diseases especially when combined with strong anti-inflammatory medications that have ulcerogenic side effects such as NSAIDs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carrageenan, suitable for gel preparation
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Indomethacin, European Pharmacopoeia (EP) Reference Standard