Merck
Narrowing the margins of errors intrinsic to the estimations made from uACR and uPCR by introducing simple correction factors.
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia (2013-07-03)
Nasrulla Abutaleb
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Creatinine, anhydrous, ≥98%