Merck
Sustain and address discharge characteristics of AC-PDP with MgO surface coated by MgO nano crystal powders.
Journal of nanoscience and nanotechnology (2013-07-19)
J H Kim, C S Park, H D Park, H S Tae, S H Lee
ABSTRAKT
This paper examined the sustain and address discharge characteristics of ac-PDPs with MgO surface coated by MgO nano crystal powders. The MgO nano crystal powder was deposited by about 5% on the MgO surface by using the spray method. To investigate the effects of the partial addition of MgO nano crystal powders on the sustain discharge as well as the address discharge, the luminance, IR spectra of 823, 828 nm, cathodoluminance, and firing voltage were measured with the measurement of the address delay times and wall voltage variation in the 42-inch ac-PDP module with a high Xe content of 17%. As a result, the statistical delay characteristics were improved considerably especially under the low panel temperature of -5 degrees C for the MgO surface with MgO nano crystal powder. However, both MgO surfaces with and without the MgO nano crystal powder showed almost similar sustain and address discharge characteristics except the statistical delay characteristics.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy