Merck
Rocuronium anaphylaxis in a 3-year-old girl with no previous exposure to neuromuscular blocking agents.
Asian Pacific journal of allergy and immunology (2013-07-19)
Sun-Hee Choi, Jae-Woo Yi, Yeong-Ho Rha
ABSTRAKT
During the perioperative period, anaphylactic reactions rarely occur. Neuromuscular blocking agents (NMBAs) are responsible for 60-70% of perioperative anaphylactic reactions. This case, we report a case of rocuronium-induced anaphylaxis in a 3-year-old girl with no previous exposure to NMBAs. This case cautions and informs practitioners that an IgE-mediated anaphylactic reaction with rocuronium is possible even in young children with no previous exposure to NMBAs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rocuronium bromide, ≥97% (perchloric acid titration)
Rocuronium for peak identification, European Pharmacopoeia (EP) Reference Standard