Merck
Platelet inhibition with cangrelor during PCI.
The New England journal of medicine (2013-07-26)
Deepak L Bhatt, Robert A Harrington
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Ticlopidine hydrochloride, analytical standard, for drug analysis
Ticlopidine hydrochloride, European Pharmacopoeia (EP) Reference Standard