Merck
  • Effect of transdermal testosterone or alpha-lipoic acid on erectile dysfunction and quality of life in patients with type 2 diabetes mellitus.
Effect of transdermal testosterone or alpha-lipoic acid on erectile dysfunction and quality of life in patients with type 2 diabetes mellitus.
Folia medica (2013-08-03)
Mitko D Mitkov, Ivanka Y Aleksandrova, Maria M Orbetzova
ABSTRAKT
Erectile dysfunction (ED) is the inability to develop and/or maintain an erection that is sufficient for satisfactory sexual intercourse. The prevalence of erectile dysfunction in diabetic men is 28-75%, this percentage rising with patient's age and duration of diabetes. The AIM of the present study was to investigate erectile dysfunction and quality of life in patients with type 2 diabetes mellitus (T2DM) after treating them with transdermal testosterone or with alpha-lipoic acid. The effect of a 12-week treatment with transdermal testosterone or alpha-lipoic acid on the erectile function and quality of life of 45 men with ED and T2DM was studied in a randomized, prospective, open clinical, comparative study. The parameters we measured in the patients were body weight and body mass index (BMI); the albumin, lipids, HbA1C, testosterone (T), sex hormone-binding globulin (SHBG), follicle stimulating hormone (FSH), luteinizing hormone (LH) and microalbuminuria levels; the International Index of Erectile Function (IIEF) and Health related quality of life (SF-36) questionnaires were completed to evaluate ED and quality of life before and after 12 weeks of treatment with alpha-lipoic acid (600 mg, parenterally, for 7 days, followed by 600 mg received per os) or with transdermal testosterone in a dose of 50 mg daily. Testosterone treatment decreased BMI significantly (p < 0.01), increased testosterone concentrations (p < 0.01) and raised the SHBG levels (p < 0.05), improved the glycemic control and lipid profile (total cholesterol, p < 0.05; HDL cholesterol, p < 0.05; triglycerides, p < 0.05). The patients treated with alpha-lipoic acid had their BMI (p < 0.01), HbA1C (p < 0.01), total cholesterol (p < 0.01), HDL-cholesterol (p < 0.01) and triglycerides (p < 0.01) significantly reduced. The indicators for ED in both groups were also statistically significantly improved. There was improvement for all patients' self-assessment score for "physical functioning" (p = 0.001), for "role limitations due to physical health" (p < 0.001) and for "general health perception" (p = 0.021). Transdermal testosterone and alpha-lipoic acid have a tangible beneficial effect on erectile dysfunction and on metabolic disorders in T2DM patients and can be used to treat such patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Testosterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®