Merck
Cardiovascular effects induced by linalool in normotensive and hypertensive rats.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2013-08-09)
Paulo J C Anjos, Aline O Lima, Patrícia S Cunha, Damião P De Sousa, Alexandre S C Onofre, Thais P Ribeiro, Isac A Medeiros, Angelo R Antoniolli, Lucindo J Quintans-Júnior, Márcio R V Santosa
ABSTRAKT
Linalool is a monoterpene alcohol and constituent of several Brazilian aromatic medicinal plants, popularly used against hypertension. Cardiovascular effects induced by linalool were evaluated. In normotensive rats, (+/-)-linalool [1, 5, 10, and 20 mg/kg body weight (BW); intravenous (i.v.)]-induced hypotension was associated with tachycardia, which was attenuated by atropine (2 mg/kg BW) and N(G)-nitro-L-arginine methyl ester (20 mg/kg BW), but was not modified after indomethacin (5 mg/kg BW) administration. In hypertensive rats, linalool [200 mg/kg BW; oral (v.o.)] reduced blood pressure without changing the heart rate. In intact rings of rat mesenteric artery precontracted with 10 microM phenylephrine, linalool (from 6.4 x 10(-6) to 6.4 x 10(-3) M) induced relaxations in a concentration-dependent manner [E(max) = (115 +/- 13)%] that were not changed after atropine administration [E(max) = (105 +/- 2)%], and were not different from those obtained in endothelium-denuded rings precontracted with phenylephrine [E(max) = (108 +/- 7)%] or 80 mM KCl [E(max) = (113 +/- 7)%] or tetraethylammonium incubation [E(max) = (105 +/- 12)%]. Linalool (1.9 x 10(-3) M) antagonized the contractions induced by CaCl2 (3 x 10(-6)-10(-2) M) (maximal inhibition, 81%). Furthermore, linalool inhibited the contractions induced by 10 microM phenylephrine or 20 mM caffeine. In conclusion, these results demonstrate that linalool reduces blood pressure probably due to a direct effect on the vascular smooth muscle leading to vasodilation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Linalool, 97%
Supelco
Linalool, analytical standard
Sigma-Aldrich
L-Linalool, natural, ≥95%, FG
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Supelco
(−)-Linalool, analytical standard