Merck
Amino-functionalized core-shell magnetic mesoporous composite microspheres for Pb(II) and Cd(II) removal.
Journal of environmental sciences (China) (2013-08-09)
Yulin Tang, Song Liang, Juntao Wang, Shuili Yu, Yilong Wang
ABSTRAKT
Amino-functionalized Fe3O4@mesoporous SiO2 core-shell composite microspheres NH2-MS in created in multiple synthesis steps have been investigated for Pb(II) and Cd(II) adsorption. The microspheres were characterized by transmission electron microscope (TEM), scanning electron microscope (SEM), N2 adsorption-desorption, zeta potential measurements and vibrating sample magnetometer. Batch adsorption tests indicated that NH2-MS exhibited higher adsorption affinity toward Pb(II) and Cd(II) than MS did. The Langmuir model could fit the adsorption isotherm very well with maximum adsorption capacity of 128.21 and 51.81 mg/g for Pb(II) and Cd(II), respectively, implying that adsorption processes involved monolayer adsorption. Pb(II) and Cd(II) adsorption could be well described by the pseudo second-order kinetics model, and was found to be strongly dependent on pH and humic acid. The Pb(II)- and Cd(II)-loaded microspheres were effectively desorbed using 0.01 mol/L HCl or EDTA solution. NH2-MS have promise for use as adsorbents in the removal of Pb(II) and Cd(II) in wastewater treatment processes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis