Merck
[The reaction of the endocrine cells of the gastrointestinal tract in response to exposure to 3,6-dichloropicolinic acid].
Biulleten' eksperimental'noi biologii i meditsiny (1989-06-01)
V V Iaglov, Iu R Ptashekas
ABSTRAKT
Silver-impregnated argentaffin cells both Ec, D1, G, P, S and K enteroendocrine cell types of rat gastrointestinal mucosa were investigated under light and electron microscopy after single oral administration of 3,6-dichloropicolinic acid. As a result, in pyloric-duodenal region Ec cells after a period of 1 hour had intensively secreted hormones. Ultrastructurally their cytoplasm and secretory granules were covered by expressed vacuolization. These data hypothetically elucidate the role of the endocrine regulations in developing barrier function of the gastrointestinal epithelium.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Clopyralid, PESTANAL®, analytical standard