Merck
Occupational dermatitis to dihydroxydiphenyl and diphenylthiourea in neoprene gloves.
Contact dermatitis (1987-05-01)
M L Masmoudi, J M Lachapelle
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4,4′-Dihydroxybiphenyl, 97%
Sigma-Aldrich
N,N′-Diphenylthiourea, 98%