Merck
Late onset isofenphos neurotoxicity.
Journal of neurology, neurosurgery, and psychiatry (1988-10-01)
A Catz, B Chen, I Jutrin, L Mendelson
ABSTRAKT
Late progressive polyneuropathy followed by pyramidal findings in a 20 year old agricultural labourer who ingested Isofenphos solution during his work, is presented. The patient was confined to a wheelchair within 6 weeks, regained walking ability within 6 months, and a 23 months' follow up revealed slight additional clinical improvement with minimal progression of the pyramidal signs. The neuropathic clinical manifestations, the EMG and the nerve conduction studies were compatible with a pathology of a distal, mainly axonal, mainly motor neuropathy.